Supelco® Organic Reference Standard, (S)-3-Hydroxybutyric acid
Supplier: Merck
Certificates
About this item
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338
Specifications
- CAS No.:6168-83-8
- Storage conditions:2…8 °C
- Hazard class description:H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD00137685
- Molecular weight:104,10
Specifications
Frequently Bought Together