Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cy3B NHS ester

Cy3B NHS ester

5 mg
Catalog # AATB949
Supplier:   AAT Bioquest
Cy3B NHS ester
Catalog # AATB949
Supplier:  AAT Bioquest
CAS Number:  
Cy3B NHS ester

5 mg

CAS Number:  
Price
2.980,00 kr
Log in for availability
1 * 5 mg
Product Details & Documents

Cy3® dye is one of the most common cyanine dyes used for labeling proteins, nucleic acids and other biological molecules.

  • Stable for 12 months

Quality tested in-house

Caution: For Research Use Only

  • Molecular Weight:657.74 g/mol
Specifications

Specifications

  • Pk:
    5 mg

Specifications