Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Amylite™ Red
Amylite™ Red
Amylite™ Red
Amylite™ Red
Catalog # AATB23040
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
Amylite™ Red
Catalog # AATB23040
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Pk:
100 Tests
Seq:
0790
Specifications
About this item
Product Details
:
:
391.23 g/mol
Product Details