Order Entry
Denmark
Orders LinkContactUsLinkComponent
5-CFDA AM
5-CFDA AM
1 mg
Catalog # AATB21910
Supplier:   AAT Bioquest
5-CFDA AM
Catalog # AATB21910
Supplier:  AAT Bioquest
CAS Number:  
5-CFDA AM
1 mg
CAS Number:  
Price
1.000,00 kr
Log in for availability
1 * 1 mg
Product Details & Documents
5-Carboxyfluorescein diacetate, acetoxymethyl ester (CFDA, AM) is a cell-permeant esterase substrate that can serve as a viability probe that measures both enzymatic activity, which is required to activate its fluorescence, and cell-membrane integrity, which is required for intracellular retention of their fluorescent product.
  • Stable for 12 months
Quality tested in-house
Specifications
Specifications
  • Pk:
    1 mg
Specifications