Anti-CD69 Mouse Monoclonal Antibody [Clone: FN50] (FITC (Fluorescein Isothiocyanate))
ANTIA86794-100
New Product
- Antibody type:Primary
- Antigen name:Activation inducer molecule
- Antigen symbol:CD69
- Clonality:Monoclonal
- Clone:FN50
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q07108
- Antigen synonyms:AIM|Activation inducer molecule (AIM/CD69)|CD69 molecule|BLAC/P26|C-type lectin domain family 2 member C|CD69 antigen (p60, early T-cell activation antigen)|CD69 antigen|CD69|BL-AC/P26
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Anti-µ-stimulated human B lymphocytes
- Purification:This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Mouse monoclonal [FN50] antibody to CD69 (FITC) for Flow Cytometry with samples derived from Human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: FN50
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human