Order Entry
Denmark
ContactUsLinkComponent
 
Anti-Galectin 3 Rabbit Recombinant Antibody [clone: LGALS3/7036R]
  ANTIA278609-100
New Product
undefined
Anti-Galectin 3 Rabbit Recombinant Antibody [clone: LGALS3/7036R]
  ANTIA278609-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    35 kDa lectin
  • Antigen symbol:
    Galectin 3
  • Clonality:
    Recombinant
  • Clone:
    LGALS3/7036R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P17931
  • Antigen synonyms:
    Carbohydrate-binding protein 35|Galactose-specific lectin 3|GAL3|CBP35|GALBP|CBP 35|Carbohydrate binding protein 35|Gal-3|Galactoside-binding protein
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    26 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to the C terminus, around amino acids 150-200, of human Galectin 3 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant rabbit monoclonal [LGALS3/7036R] antibody to Galectin 3 for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Galectin 3
Clonality: Recombinant
Clone: LGALS3/7036R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human