Anti-HIC2 Mouse Monoclonal Antibody [clone: PCRP-HIC2-2F8]
ANTIA277609-100
New Product
- Antibody type:Primary
- Antigen name:HIC1 related gene on chromosome 22
- Antigen symbol:HIC2
- Clonality:Monoclonal
- Clone:PCRP-HIC2-2F8
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q96JB3
- Antigen synonyms:Hic3|KIAA1020|ZNF907|ZBTB30|HRG22|HIC2|Hypermethylated in cancer 2 protein|Zinc finger and BTB domain-containing protein 30
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:66,1 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human HIC2 protein.
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [PCRP-HIC2-2F8] antibody to HIC2 for ELISA, IP, Flow Cytometry and IF with samples derived from human.
- Validated Applications: ELISA, IP, Flow Cytometry, IF
- Recommended Dilutions: IP: 1 to 2µg / 100 to 500µg proteins, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: HIC2
Clonality: Monoclonal
Clone: PCRP-HIC2-2F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human