Anti-TRP1 Rabbit Recombinant Antibody [clone: TYRP1/2340R]
Catalog # ANTIA253439-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Glycoprotein 75
- Antigen symbol:TRP1
- Clonality:Recombinant
- Clone:TYRP1/2340R
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P17643
- Antigen synonyms:CAS2|Catalase B|6-dihydroxyindole-2-carboxylic acid oxidase|GP75|CATB|b-PROTEIN|5,6 dihydroxyindole 2 carboxylic acid oxidase|DHICA oxidase|5
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:75 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, within amino acids 257 - 377, of human TYRP1 protein. The exact sequence is proprietary
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [TYRP1/2340R] antibody to TRP1 for ELISA and IHC-P with samples derived from human.
- Validated Applications: ELISA, IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: TRP1
Clonality: Recombinant
Clone: TYRP1/2340R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human