Order Entry
Denmark
ContactUsLinkComponent
 
Anti-Mucin 5AC Rabbit Recombinant Antibody [Clone: MUC5AC/7067R]
  ANTIA278620-100
New Product
undefined
Anti-Mucin 5AC Rabbit Recombinant Antibody [Clone: MUC5AC/7067R]
  ANTIA278620-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Apomucin
  • Antigen symbol:
    Mucin 5AC
  • Clonality:
    Recombinant
  • Clone:
    MUC5AC/7067R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P98088
  • Antigen synonyms:
    Lewis B blood group antigen|Gastric mucin|Mucin 5 Subtypes A And C|MUC5AC|Apomucin Major Airway Glycoprotein|MUC 5|MUC5|Mucin 5 subtype AC tracheobronchial|LeB
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    >1,000 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human MUC5AC protein.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant rabbit monoclonal [MUC5AC/7067R] antibody to Mucin 5AC for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Mucin 5AC
Clonality: Recombinant
Clone: MUC5AC/7067R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human