Anti-ZBED1 Mouse Monoclonal Antibody [Clone: PCRP-ZBED1-1E1]
Catalog # ANTIA253690-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Ac like transposable element
- Antigen symbol:ZBED1
- Clonality:Monoclonal
- Clone:PCRP-ZBED1-1E1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# O96006
- Antigen synonyms:dREF homolog|ALTE|BED type zinc finger domain containing protein 1|Putative Ac like transposon|Putative Ac-like transposable element|KIAA0785|DREF|hDREF|DNA replication related element binding factor
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:86 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human ZBED1 protein.
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [PCRP-ZBED1-1E1] antibody to ZBED1 for Flow Cytometry, IF and WB with samples derived from human.
- Validated Applications: Flow Cytometry, IF, WB
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml
Type: Primary
Antigen: ZBED1
Clonality: Monoclonal
Clone: PCRP-ZBED1-1E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human