Anti-SUMO-1 Mouse Monoclonal Antibody [clone: SUMO1/1188]
ANTIA253448-100
New Product
- Antibody type:Primary
- Antigen name:DAP1
- Antigen symbol:SUMO-1
- Clonality:Monoclonal
- Clone:SUMO1/1188
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P63165
- Antigen synonyms:Sentrin|GMP 1|GAP-modifying protein 1|GAP modifying protein 1|PIC 1|GMP1|OFC10|PIC1|SENP2
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:11,5 kDa (monomer)/90 kDa (heteromer)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human SUMO1 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [SUMO1/1188] antibody to SUMO-1 for Flow Cytometry, IF, WB and IHC-P with samples derived from human and rat.
- Validated Applications: Flow Cytometry, IF, WB, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: SUMO-1
Clonality: Monoclonal
Clone: SUMO1/1188
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat