Anti-Collagen IV Rabbit Recombinant Antibody [clone: COL4/8657R]
Catalog # ANTIA314808-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Arresten
- Antigen symbol:Collagen IV
- Clonality:Recombinant
- Clone:COL4/8657R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P02462, P08572, P29400, P53420
- Antigen synonyms:RATOR|collagen alpha-1(IV) chain|Collagen IV|CO4A1_HUMAN|BSVD|COL4A1|collagen type IV alpha 1 chain
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:161 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Purified human Collagen IV protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [COL4/8657R] antibody to Collagen IV for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Collagen IV
Clonality: Recombinant
Clone: COL4/8657R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human