Order Entry
Denmark
ContactUsLinkComponent
 
Anti-ATF1 Goat Polyclonal Antibody
  ANTIA85122-100
New Product
undefined
Anti-ATF1 Goat Polyclonal Antibody
  ANTIA85122-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Activating transcription factor 1
  • Antigen symbol:
    ATF1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    cAMP dependent transcription factor 1|ATF1_HUMAN|ATF1 FUS fusion gene|ATF1|cAMP-dependent transcription factor ATF-1|ATF1 EWS fusion gene|Cyclic AMP dependent transcription factor ATF1|Cyclic AMP dependent transcription factor ATF 1|ATF 1
  • Storage buffer:
    Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
  • Sequence:
    EDSHKSTTSETAPQ-C
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    100 µg at 0,5 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to Human ATF1 (N terminal)
  • Purification:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Goat polyclonal antibody to ATF1 for ELISA and WB with samples derived from human.

  • Validated applications: ELISA, W

Type: Primary
Antigen: ATF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human