Anti-AMPK alpha 2 Goat Polyclonal Antibody
Catalog # ANTIA84859-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:5'-AMP-activated protein kinase catalytic subunit alpha-2
- Antigen symbol:AMPK alpha 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:ACACA kinase|AMPKa2|AMPK alpha 2|AMPKalpha2|AMPK2|AMPK subunit alpha-2|Acetyl-CoA carboxylase kinase|AMPK alpha 2 chain|AAPK2_HUMAN
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:CPLDALNTTKP
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human PRKAA2 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to AMPK alpha 2 for ELISA and WB with samples derived from human, mouse and rat.
- Validated applications: ELISA, W
Type: Primary
Antigen: AMPK alpha 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse;Rat