Anti-CREB3L2 Goat Polyclonal Antibody
ANTIA84748-100
New Product
- Antistof type:Primary
- Antigen name:B ZIB transcription factor
- Antigen symbol:CREB3L2
- Clonality:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Vært:Goat
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonymer:cAMP responsive element binding protein 3 like 2|BBF2H7|BZIB transcription factor|cAMP-responsive element-binding protein 3-like protein 2|Basic transcription factor 2|Cyclic AMP-responsive element-binding protein 3-like protein 2|BBF2 human homolog on chromosome 7|CR3L2_HUMAN|CREB3L2
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sekvens:C-HSLQEPYTASVVRS
- Opbevaringstemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Koncentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human CREB3L2 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Pakning:Plastic vial
- Pakn:100 µG
Goat polyclonal antibody to CREB3L2 for ELISA and WB with samples derived from mouse and rat.
- Validated applications: ELISA, W
Type: Primary
Antigen: CREB3L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse;Rat