Order Entry
Denmark
ContactUsLinkComponent
Anti-CREB3L1 Goat Polyclonal Antibody
Anti-CREB3L1 Goat Polyclonal Antibody
Catalog # ANTIA84126-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-CREB3L1 Goat Polyclonal Antibody
Catalog # ANTIA84126-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    BBF-2 homolog
  • Antigen symbol:
    CREB3L1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    Processed cyclic AMP-responsive element-binding protein 3-like protein 1|Old astrocyte specifically-induced substance|OASIS|CAMP responsive element binding protein 3 like 1|Cyclic AMP responsive element binding protein 3 like protein 1|CR3L1_HUMAN|cAMP-responsive element-binding protein 3-like protein 1|CREB3L1
  • Storage buffer:
    Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
  • Sequence:
    C-DFSHSKEWFHDRD
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    100 µg at 0,5 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to Human CREB3L1 (C terminal)
  • Tested applications:
    IHC
  • Purification:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Goat polyclonal antibody to CREB3L1 for ELISA, WB and IHC with samples derived from human.

  • Validated applications: ELISA, WB, IH

Type: Primary
Antigen: CREB3L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human