Anti-UGT8 Rabbit Polyclonal Antibody
Catalog # ANTIA99847-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:2 hydroxyacylsphingosine 1 beta galactosyltransferase
- Antigen symbol:UGT8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q16880
- Antigen synonyms:CGT|UDP galactose ceramide galactosyltransferase|mCerGT|Cerebroside synthase|2-hydroxyacylsphingosine 1-beta-galactosyltransferase|CGT_HUMAN|Ceramide UDP-galactosyltransferase|Ceramide UDP galactosyltransferase|UDP galactose ceramide galactosyltransferase 8
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg⁺² and Ca⁺²), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:61 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human UGT8 (amino acids 492-541)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to UGT8 for WB and ELISA with samples derived from human and rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: UGT8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat