Anti-DAK Rabbit Polyclonal Antibody
ANTIA98531-100
New Product
- Antibody type:Primary
- Antigen name:ATP dependent dihydroxyacetone kinase
- Antigen symbol:DAK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q3LXA3
- Antigen synonyms:Dha kinase/FMN cyclase|Bifunctional ATP dependent dihydroxyacetone kinase/FAD AMP lyase (cyclizing)|DHA kinase|FAD AMP lyase (cyclic FMN forming)|Dihydroxyacetone kinase 2 homolog (S. cerevisiae)|DHAK_HUMAN|DAK|Dihydroxyacetone kinase 2|Dihydroxyacetone kinase 2 homolog (yeast)
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:58 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human DAK (amino acids 91-140)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to DAK for WB and ELISA with samples derived from human, mouse and rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000
Type: Primary
Antigen: DAK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat