Anti-ATF1 Rabbit Polyclonal Antibody
Plastic vial
Catalog # ANTIA98282-100
Supplier: Antibodies.com
Jump to:
Product Details & Documents
Rabbit polyclonal antibody to ATF1 for WB, IHC and ELISA with samples derived from Human and Mouse.
Recommended dilutions: WB: 1:500 to 1:1000 IHC: 1:50 to 1:100 ELISA: 1:1000 .
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Type: Primary
Antigen: ATF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Seq:1286
- Pack type:Plastic vial
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Host:Rabbit
- Molecular weight:29 kDa
- Shipping temperature:Shipped on blue ice at 4 °C
- Antigen name:Activating transcription factor 1
- Cat. No.:ANTIA98282-100
- Reactivity:Human|Mouse
- Gene ID:UniprotID# P18846
- Antigen synonyms:Activating transcription factor 1|cAMP-dependent transcription factor ATF-1|Cyclic AMP-dependent transcription factor ATF-1|Protein TREB36
- Immunogen:Synthetic peptide derived from human ATF1 (amino acids 31-80)
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Tested applications:IHC
- Antigen symbol:ATF1
- Storage buffer:Supplied in pH osphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150mM NaCl, 0,02% Sodium Azide, and 50% Glycerol.
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



