Order Entry
Denmark
ContactUsLinkComponent
Anti-Protein C Rabbit Monoclonal Antibody [clone: ARC1023]
Anti-Protein C Rabbit Monoclonal Antibody [clone: ARC1023]
Catalog # ANTIA306260-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Protein C Rabbit Monoclonal Antibody [clone: ARC1023]
Catalog # ANTIA306260-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Activation peptide
  • Antigen symbol:
    Protein C
  • Clonality:
    Monoclonal
  • Clone:
    ARC1023
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P04070
  • Antigen synonyms:
    proC|APC|PROC_HUMAN|EC 3.4.21.69|Autoprothrombin IIA|Anticoagulant protein C|PROC1|Protein C (inactivator of coagulation factors Va and VIIIa)|Blood coagulation factor XIV
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
  • Molecular weight:
    52 kDa
  • Sequence:
    IALLHLAQPATLSQTIVPICLPDSGLAERELNQAGQETLVTGWGYHSSREKEAKRNRTFVLNFIKIPVVPHNECSEVMSNMVSENMLCAGILGDRQDACEGDSGGPMVASFHGTWFLVGLVSWGEGCGLLHNYGVYTKVSRYLDWIHGHIRDKEAPQKSWAP
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 300-461 of human Coagulation Coagulation Protein C (P04070).
  • Tested applications:
    ICC/IF
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit monoclonal [ARC1023] antibody to Protein C for WB and ICC/IF with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, ICC/IF
  • Recommended Dilutions: WB: 1:500 to 1:2000, ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: Protein C
Clonality: Monoclonal
Clone: ARC1023
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat