Anti-AK3L1 Rabbit Polyclonal Antibody
ANTIA307027-100
New Product
- Antibody type:Primary
- Antigen name:Adenylate kinase 3 like 1
- Antigen symbol:AK3L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P27144
- Antigen synonyms:GTP:AMP phosphotransferase|adenylate kinase 4|Adenylate kinase isoenzyme 4, mitochondrial|AK3|AK4|AK3L1|ATP-AMP transphosphorylase|Adenylate kinase 3-like|AK3L2
- Storage buffer:Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% Thiomersal
- Molecular weight:25 kDa
- Sequence:MASKLLRAVILGPPGSGKGTVCQRIAQNFGLQHLSSGHFLRENIKASTEVGEMAKQYIEKSLLVPDHVITRLMMSELENRRGQHWLLDGFPRTLGQAEALDKICEVDLVISLNIPFETLKDRLSRRWIHPPSGRVYNLDFNPPHVHGIDDVTGEPLVQQEDDKPEAVAARLRQYKDVAKPVIELYKSRGVLHQFSGTETNKIWPYVYTLFSNKITPIQSKEAY
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-223 of human AK4 (NP_037542.1).
- Tested applications:ICC/IF
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Rabbit polyclonal antibody to AK3L1 for WB, ICC/IF and IP with samples derived from Human and Mouse.
- Validated Applications: WB, ICC/IF, IP
- Recommended Dilutions: WB: 1:500 to 1:2000, ICC/IF: 1:50 to 1:200, IP: 1:50 to 1:200
Type: Primary
Antigen: AK3L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse