Order Entry
Denmark
ContactUsLinkComponent
iFluor® 488-dUTP *1 mM in TE Buffer (pH 7,5)*
iFluor® 488-dUTP *1 mM in TE Buffer (pH 7,5)*
Catalog # AATB17044
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
undefined
iFluor® 488-dUTP *1 mM in TE Buffer (pH 7,5)*
Catalog # AATB17044
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    100 Nmo
Specifications
About this item
This iFluor 488-dUTP conjugate is used as a green fluorescence colour with SpectrumGreen™ filter set (SpectrumGreen™ is the trademark of Vysis).
  • Widely used for both FISH probes and microarray-based experiments
  • Bright signal that has high photostability and is not affected by pH
The dye-modified deoxyuridine 5'-triphosphates are one of the most common methods to produce dye-labelled DNA via the conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labelling or PCR.
Caution: For research use only.