Anti-Mucin 5AC Rabbit Recombinant Antibody [clone: MUC5AC/8340R]
Catalog # ANTIA316726-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Apomucin
- Antigen symbol:Mucin 5AC
- Clonality:Recombinant
- Clone:MUC5AC/8340R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P98088
- Antigen synonyms:Lewis B blood group antigen|Gastric mucin|Mucin 5 Subtypes A And C|MUC5AC|Apomucin Major Airway Glycoprotein|MUC 5|MUC5|Mucin 5 subtype AC tracheobronchial|LeB
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:>1000 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant full-length human MUC5AC protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [MUC5AC/8340R] antibody to Mucin 5AC for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Mucin 5AC
Clonality: Recombinant
Clone: MUC5AC/8340R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human