Anti-Mucin 5AC Rabbit Recombinant Antibody [clone: MUC5AC/8340R]
ANTIA315342-100
New Product
- Antistof type:Primary
- Antigen name:Apomucin
- Antigen symbol:Mucin 5AC
- Clonality:Recombinant
- Clone:MUC5AC/8340R
- Konjugation:Unconjugated
- Vært:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gen ID:UniprotID# P98088
- Antigen synonymer:Lewis B blood group antigen|Gastric mucin|Mucin 5 Subtypes A And C|MUC5AC|Apomucin Major Airway Glycoprotein|MUC 5|MUC5|Mucin 5 subtype AC tracheobronchial|LeB
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:>1000 kDa
- Opbevaringstemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Koncentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human MUC5AC protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pakning:Plastic vial
- Pakn:100 µG
Recombinant rabbit monoclonal [MUC5AC/8340R] antibody to Mucin 5AC for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Mucin 5AC
Clonality: Recombinant
Clone: MUC5AC/8340R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human