Anti-MBD1 Mouse Monoclonal Antibody [clone: CPTC-MBD1-1]
Catalog # ANTIA249280-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CXXC 3
- Antigen symbol:MBD1
- Clonality:Monoclonal
- Clone:CPTC-MBD1-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q9UIS9
- Antigen synonyms:CXXC3|MBD 1|MBD1_HUMAN|CXXC-type zinc finger protein 3|Methyl CpG binding domain protein 1 isoform PCM1|MBD1|Methyl CpG binding domain protein 1|MECP1 COMPLEX|Methyl CpG binding protein MBD1
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:80 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human MBD1 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [CPTC-MBD1-1] antibody to MBD1 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: MBD1
Clonality: Monoclonal
Clone: CPTC-MBD1-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human