Order Entry
Denmark
ContactUsLinkComponent
iFluor® 488 PEG12-dUTP *1 mM in TE Buffer (pH 7,5)*
iFluor® 488 PEG12-dUTP *1 mM in TE Buffer (pH 7,5)*
Catalog # AATB17041
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
undefined
iFluor® 488 PEG12-dUTP *1 mM in TE Buffer (pH 7,5)*
Catalog # AATB17041
Supplier:  AAT BIOQUEST
New Product
CAS Number:  

Specifications

  • Pk:
    25 Nmo
  • Application:
  • Environmentally Preferable:

Specifications

About this item

This iFluor® 488-PEG12-dUTP conjugate has the same spectral properties to ChromaTide™ Alexa Fluor 488-dUTP.

  • PEG12 spacer makes it much more effectively incorporated into DNAs by polymerase
  • Bright signal that has high photostability and is not affected by pH

The dye-modified deoxyuridine 5'-triphosphates are one of the most common methods to produce dye-labelled DNA via the conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labelling or PCR.

Caution: For research use only.