Order Entry
Denmark
Orders LinkContactUsLinkComponent
V.C.N. Supplement

V.C.N. Supplement

V.C.N. Supplement, 5×500 ml, 5 Vial
Catalog # HIMEFD023-5X5VL
Supplier:   HIMEDIA
V.C.N. Supplement
Catalog # HIMEFD023-5X5VL
Supplier:  HIMEDIA
CAS Number:  
V.C.N. Supplement

V.C.N. Supplement, 5×500 ml, 5 Vial

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 Each
Product Details & Documents

An antibiotic supplement, recommended for the selective isolation of N. gonorrhoeae and N. meningitidis.

  • Store at 2 to 8 °C
Specifications

Specifications

  • Pk:
    5 Vial
  • Description:
    V.C.N. Supplement, 5×500 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.