Order Entry
Denmark
ContactUsLinkComponent
Anti-CD123 (IL-3Rα) Mouse Monoclonal Antibody [clone: 6H6] (APC)
Anti-CD123 (IL-3Rα) Mouse Monoclonal Antibody [clone: 6H6] (APC)
Certificates
undefined
Anti-CD123 (IL-3Rα) Mouse Monoclonal Antibody [clone: 6H6] (APC)

Specifications

  • Catalog No:
  • STMC60110AZ
  • STMC60110AZ.1
  • Antigen name:
  • CD123 (IL-3Ra)
  • CD123 (IL-3Ra)
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • 6H6
  • 6H6
  • Reactivity:
  • Sooty mangabey, Rhesus monkey, Human
  • Sooty mangabey, Rhesus monkey, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3563
  • 3563
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together