About this item
Mouse monoclonal IgG2b antibody against human CD68, Alexa Fluor® 488-conjugated.
- Granulocytes
- Subsets
- Myeloid cells
The Y1/82A antibody reacts with human CD68, an ~110 kDa type 1 transmembrane glycoprotein and member of the sialomucin, LAMP and scavenger receptor families. The heavily glycosylated extracellular domain of CD68, which contains a praline-rich spacer region separating mucin and LAMP domains, binds low-density lipoprotein and certain lectins and selectins, though the function of CD68 remains unclear. CD68 is highly expressed in lysosomes, endosomes and cytoplasmic granules, and more weakly on the surface of macrophages, monocytes, neutrophils, basophils, dendritic cells and NK cells. It has also been detected in the cytoplasm of γ/δ T cells, LAK cells, fibroblasts, endothelial cells and subsets of B cells and hematopoietic progenitors, as well as in various non-hematopoietic tissues such as liver and kidney. The Y1/82A antibody recognizes an epitope distinct from those of antibody clones Y2/131, EBM11, Ki-M6 and KP1. Y1/82A is reportedly more specific for monocytes and macrophages than KP1.
Type:
Antigen: CD68
Clonality: Monoclonal
Clone: Y1/82A
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Specifications
- Catalog No:
- STMC60105AD.1
- STMC60105AD
- Antigen name:
- CD68
- CD68
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Alexa Fluor® 488
- Alexa Fluor® 488
- Clone:
- Y1/82A
- Y1/82A
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 968
- 968
- Isotype:
- IgG2b kappa
- IgG2b kappa