Order Entry
Denmark
ContactUsLinkComponent
Anti-Igg1 Isotype Control Rat Monoclonal Antibody [clone: RTK2071] (FITC)
Anti-Igg1 Isotype Control Rat Monoclonal Antibody [clone: RTK2071] (FITC)
Certificates
undefined
Anti-Igg1 Isotype Control Rat Monoclonal Antibody [clone: RTK2071] (FITC)

Specifications

  • Catalog No:
  • STMC60075FI.1
  • STMC60075FI
  • Antigen name:
  • Igg1 Isotype Control
  • Igg1 Isotype Control
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • RTK2071
  • RTK2071
  • Host:
  • Rat
  • Rat
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together