Anti-CD43 Rat Monoclonal Antibody [clone: R2/60] (Biotin)
Supplier: STEMCELL Technologies
Certificates
About this item
Rat monoclonal IgM antibody against mouse CD43, biotin-conjugated.
- B cells
- Dendritic cells
- Granulocytes
- Subsets
- Hematopoietic Stem
- Progenitor cells
- Macrophages
- Monocytes
- Myeloid cells
- Nk cells
- Plasma
- Platelets
- T cells
The R2/60 antibody reacts with mouse CD43, a type I transmembrane sialoglycoprotein expressed on the surface of most hematopoietic cells including thymocytes, T cells, B cells, NK cells, monocytes, granulocytes, platelets, and hematopoietic progenitor cells. CD43 binds ligands such as CD54 and CD169 (siglec-1) and plays regulatory roles in cell adhesion, activation, and survival. CD43 exhibits two different glycoforms; a 115 kDa form, and an activation-associated 130 kDa form, both of which are detected by the R2/60 antibody.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Type:
Antigen: CD43
Clonality: Monoclonal
Clone: R2/60
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgM kappa
Reactivity: Mouse
Specifications
- Catalog No:
- STMC60042BT.1
- STMC60042BT
- Antigen name:
- CD43
- CD43
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- R2/60
- R2/60
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 20737
- 20737
- Isotype:
- IgM kappa
- IgM kappa
Specifications
Frequently Bought Together