Order Entry
Denmark
ContactUsLinkComponent
 
Anti-CD14 Mouse Monoclonal Antibody [clone: M5E2] (PE)
 
undefined
Anti-CD14 Mouse Monoclonal Antibody [clone: M5E2] (PE)

 

  • Catalog No:
  • STMC60004PE.1
  • STMC60004PE
  • Antigen name:
  • CD14
  • CD14
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • M5E2
  • M5E2
  • Reactivity:
  • Dog, Squirrel monkey, Capuchin monkey, Pig, Sheep, Cynomolgus monkey, Common marmoset, Rhesus monkey, Cotton-topped tamarin, Macaque, Chimpanzee, Human, Bovine
  • Dog, Squirrel monkey, Capuchin monkey, Pig, Sheep, Cynomolgus monkey, Common marmoset, Rhesus monkey, Cotton-topped tamarin, Macaque, Chimpanzee, Human, Bovine
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 929
  • 929
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

 

Frequently Bought Together