- Antigen name:SHIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Gene ID:3635 (human), 16331 (mouse)
- Pk:100 µl
Rabbit polyclonal antibody against human, mouse SHIP, unconjugated.
- Granulocytes
- Subsets
- Lymphocytes
- Myeloid cells
The SH2-containing inositol phosphatase (SHIP, also called SHIP1) is a 145 kDa hematopoietic-restricted protein that becomes tyrosine-phosphorylated and associated with the adaptor protein, Shc, following cytokine, growth factor, chemokine, and immunoreceptor stimulation. SHIP also hydrolyzes the critical phosphatidylinositol (PI)-3-kinase (PI3K)-generated second messenger, PI-3,4,5-P3 (PIP3), to PI-3,4-P2 1,2 and therefore acts as an important negative regulator of the PI3K pathway. This antibody reacts with mouse and human full-length 145 kDa SHIP protein.
Type:
Antigen: SHIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse