Order Entry
Denmark
ContactUsLinkComponent
Anti-HLA-DR Mouse Monoclonal Antibody [clone: L243] (APC)
Anti-HLA-DR Mouse Monoclonal Antibody [clone: L243] (APC)
Certificates
undefined
Anti-HLA-DR Mouse Monoclonal Antibody [clone: L243] (APC)

Specifications

  • Catalog No:
  • STMC100-0335
  • STMC100-0336
  • Antigen name:
  • HLA-DR
  • HLA-DR
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • L243
  • L243
  • Reactivity:
  • Baboon, African green monkey, Cynomolgus monkey, Common marmoset, Rhesus monkey, Cotton-topped tamarin, Pigtailed macaque, Chimpanzee, Human, Dog, Squirrel monkey
  • Baboon, African green monkey, Cynomolgus monkey, Common marmoset, Rhesus monkey, Cotton-topped tamarin, Pigtailed macaque, Chimpanzee, Human, Dog, Squirrel monkey
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3122, 3123
  • 3122, 3123

Specifications

Frequently Bought Together