About this item
Rat monoclonal IgG2a antibody against mouse CD267 (TACI), PE-conjugated.
- B cells
The 8F10 antibody reacts with mouse CD267 (transmembrane activator and CAML interactor, or TACI), an ~30 kDa type III transmembrane protein of the TNF receptor superfamily that is expressed primarily on the surface of maturing subsets of splenic B cells, including transition type 2 (T2) and marginal zone (MZ) B cells, as well as on plasma cells and plasmablasts. Expression of CD267 is upregulated on B cells exposed to LPS both in vitro and in vivo. CD267 plays important roles in humoral immunity by regulating B cell survival, proliferation, and maturation through its interaction with the ligands APRIL and BAFF. Ligand binding stimulates activation of several transcription factors, including NF-kappa-B, and has been shown to induce either proliferation or apoptosis, suggesting a dual role in the regulation of B cells. CD267 also appears to have an essential function in immunoglobulin isotype switching and B cell differentiation in response to thymus-independent antigens.
Type: Anti-CD267 (TACI) Rat Monoclonal Antibody [clone: 8F10] (PE)
Antigen: CD267 (TACI)
Clonality: Monoclonal
Clone: 8F10
Conjugation: PE
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Specifications
- Catalog No:
- STMC60116PE
- STMC60116PE.1
- Antigen name:
- CD267 (TACI)
- CD267 (TACI)
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- 8F10
- 8F10
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 57916
- 57916
- Isotype:
- IgG2a kappa
- IgG2a kappa