Order Entry
Denmark
ContactUsLinkComponent
Anti-CD43 Mouse Monoclonal Antibody [clone: CD43-10G7] (PE)
Anti-CD43 Mouse Monoclonal Antibody [clone: CD43-10G7] (PE)
Certificates
undefined
Anti-CD43 Mouse Monoclonal Antibody [clone: CD43-10G7] (PE)

Specifications

  • Catalog No:
  • STMC60085PE
  • STMC60085PE.1
  • Antigen name:
  • CD43
  • CD43
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • CD43-10G7
  • CD43-10G7
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 6693
  • 6693
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together