Order Entry
Denmark
ContactUsLinkComponent
Anti-SSEA-3 Rat Monoclonal Antibody [clone: MC-631] (Alexa Fluor® 488)
Anti-SSEA-3 Rat Monoclonal Antibody [clone: MC-631] (Alexa Fluor® 488)
Certificates
undefined
Anti-SSEA-3 Rat Monoclonal Antibody [clone: MC-631] (Alexa Fluor® 488)

Specifications

  • Catalog No:
  • STMC60061AD
  • STMC60061AD.1
  • Antigen name:
  • SSEA-3
  • SSEA-3
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Clone:
  • MC-631
  • MC-631
  • Reactivity:
  • Rhesus monkey, Human, Rat, Mouse
  • Rhesus monkey, Human, Rat, Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 93961
  • 93961
  • Isotype:
  • IgM kappa
  • IgM kappa

Specifications

Frequently Bought Together