Order Entry
Denmark
ContactUsLinkComponent
 
Anti-CD90 (Thy-1) Mouse Monoclonal Antibody [clone: 5E10] (PE)
 
undefined
Anti-CD90 (Thy-1) Mouse Monoclonal Antibody [clone: 5E10] (PE)

 

  • Catalog No:
  • STMC60045PE.1
  • STMC60045PE
  • Antigen name:
  • CD90 (Thy-1)
  • CD90 (Thy-1)
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • 5E10
  • 5E10
  • Reactivity:
  • Baboon, Pig, Cynomolgus monkey, Rhesus monkey, Macaque, Human, Dog
  • Baboon, Pig, Cynomolgus monkey, Rhesus monkey, Macaque, Human, Dog
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 7070
  • 7070
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

 

Frequently Bought Together