Order Entry
Denmark
ContactUsLinkComponent
Anti-CD45 Rat Monoclonal Antibody [clone: 30-F11] (PE)
Anti-CD45 Rat Monoclonal Antibody [clone: 30-F11] (PE)
Certificates
undefined
Anti-CD45 Rat Monoclonal Antibody [clone: 30-F11] (PE)

Specifications

  • Catalog No:
  • STMC60030PE.1
  • STMC60030PE
  • Antigen name:
  • CD45
  • CD45
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • 30-F11
  • 30-F11
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 19264
  • 19264
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa

Specifications

Frequently Bought Together