Order Entry
Denmark
ContactUsLinkComponent
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (FITC)
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (FITC)
Certificates
undefined
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (FITC)

Specifications

  • Catalog No:
  • STMC60005FI.1
  • STMC60005FI
  • Antigen name:
  • CD19
  • CD19
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • HIB19
  • HIB19
  • Reactivity:
  • Chimpanzee, Human
  • Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 930
  • 930
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together