Mouse monoclonal IgG2a antibody against human, rhesus, cynomolgus CD14, biotin-conjugated.
- Monocytes
The M5E2 antibody reacts with CD14, an ~53 to 55 kDa GPI-anchored transmembrane glycoprotein expressed at high levels on the surface of peripheral blood monocytes and macrophages, and at lower levels on granulocytes. An ~10-fold difference in expression levels between monocytes/macrophages and granulocytes makes CD14 a useful marker for distinguishing these cell populations. CD14 is also found on tissue macrophages, Langerhans cells and dendritic cells. CD14 functions as a high-affinity receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein and modulates LPS-dependent signal transduction during the immune response to gram-negative pathogens by acting as a co-receptor for TLR 4 and MD-2. This triggers activation of NF-kappa-B, cytokine secretion, and induction of the inflammatory response. Two soluble forms of CD14 have also been described (~48 and ~55 kDa). This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ human Myeloid positive selection kit (Catalog #18653) and EasySep™ human CD14 positive selection kit (Catalog #18058); partial blocking may be observed, as well as EasySep™ HLA whole blood CD33 positive selection kit (Catalog #18287HLA), EasySep™ HLA whole blood Myeloid positive selection kit (Catalog #18683HLA), and EasySep™ human Buffy Coat CD14 positive selection kit (Catalog #18088).
Type: Anti-CD14 Mouse Monoclonal Antibody [clone: M5E2] (APC)
Antigen: CD14
Clonality: Monoclonal
Clone: M5E2
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Capuchin monkey, Chimpanzee, Common marmoset, Cotton-topped tamarin, Bovine, Cynomolgus monkey, Dog, Human, Macaque, Pig, Rhesus monkey, Sheep, Squirrel monkey
- Catalog No:
- STMC60004AZ.1
- STMC60004AZ
- Antigen name:
- CD14
- CD14
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- M5E2
- M5E2
- Reactivity:
- Dog, Squirrel monkey, Capuchin monkey, Pig, Sheep, Cynomolgus monkey, Common marmoset, Rhesus monkey, Cotton-topped tamarin, Macaque, Chimpanzee, Human, Bovine
- Dog, Squirrel monkey, Capuchin monkey, Pig, Sheep, Cynomolgus monkey, Common marmoset, Rhesus monkey, Cotton-topped tamarin, Macaque, Chimpanzee, Human, Bovine
- Host:
- Mouse
- Mouse
- Gene ID:
- 929
- 929
- Isotype:
- IgG2a kappa
- IgG2a kappa