Order Entry
Denmark
ContactUsLinkComponent
Sulphuric acid
Sulphuric acid
Catalog # CMPLJPH493
New Product
CAS Number:  
Sulphuric acid
Catalog # CMPLJPH493
New Product
CAS Number:  

Specifications

  • Concentration:
    0,05 mol/L
  • Pk:
    1 L
  • Description:
    Sulfuric acid TS
  • Shipping conditions:
    Store under normal laboratory conditions, at temperatures between 15...25 °C
  • Shelf life:
    24 months

Specifications

About this item

Certifications: ISO 17034, 17025, 9001 certified.