Order Entry
Denmark
ContactUsLinkComponent
 
Sulfuric Acid TS
  CMPLJPH492
New Product
 :  
Sulfuric Acid TS
  CMPLJPH492
New Product
 :  

 

  • Pk:
    100 mL
  • Description:
    Sulfuric acid TS
  • Shipping conditions:
    Store under normal laboratory conditions, at temperatures between 15...25 °C
  • UN No.:
    2796
  • GHS signal:
    Danger
  • Shelf life:
    24 months

 

 

The standard is specially intended for use in testing Japanese pharmacopoeia and monographs. This solution was produced according of Japanese pharmacopoeia specifications using reagent quality components and water meeting the Japanese pharmacopoeia monograph requirement for purified water.

 : ISO 17034, 17025, 9001 certified.