Order Entry
Denmark
ContactUsLinkComponent
Anti-CD105 Mouse Monoclonal Antibody [clone: 43A3] (FITC)
Anti-CD105 Mouse Monoclonal Antibody [clone: 43A3] (FITC)
Certificates
undefined
Anti-CD105 Mouse Monoclonal Antibody [clone: 43A3] (FITC)

Specifications

  • Catalog No:
  • STMC60039FI
  • STMC60039FI.1
  • Antigen symbol:
  • CD105
  • CD105
  • Antigen name:
  • Endoglin
  • Endoglin
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • 43A3
  • 43A3
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 2022
  • 2022
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together