Order Entry
Denmark
ContactUsLinkComponent
Anti-CD49b Rat Monoclonal Antibody [clone: DX5] (APC)
Anti-CD49b Rat Monoclonal Antibody [clone: DX5] (APC)
Certificates
undefined
Anti-CD49b Rat Monoclonal Antibody [clone: DX5] (APC)

Specifications

  • Catalog No:
  • STMC60020AZ
  • STMC60020AZ.1
  • Antigen symbol:
  • CD49b
  • CD49b
  • Antigen name:
  • Integrin a2
  • Integrin a2
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • DX5
  • DX5
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 16398
  • 16398
  • Isotype:
  • IgM kappa
  • IgM kappa

Specifications

Frequently Bought Together