- Antigen name:Transferrin Receptor
- Antigen symbol:CD71
- Clonality:Monoclonal
- Clone:OKT9
- Conjugation:Biotin
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Gene ID:7037
- Pk:100 µG
Mouse monoclonal IgG1 antibody against human CD71 (transferrin receptor), biotin-conjugated.
- Erythroid cells
The OKT9 antibody reacts with CD71 (transferrin receptor), an ~180 - 190 kDa disulfide-bonded homodimer and type 2 transmembrane glycoprotein expressed at high levels on a broad range of actively proliferating cells, as well as reticulocytes, monocytes, macrophages and marrow stromal cells. Surface levels are low on resting leukocytes but are upregulated on lymphocytes, monocytes and macrophages following antigen or mitogen stimulation. Expression is lost during differentiation of reticulocytes into mature erythrocytes. CD71 mediates uptake of transferrin–iron complexes through receptor-mediated endocytosis and recycling of the apotransferrin–receptor complex to the cell surface. CD71 has been employed as a marker for evaluating erythroid precursors and for assessing disorders such as erythroid leukemia and myelodysplastic syndrome. It has also served as a target for drug delivery. The OKT9 antibody binds an epitope that reportedly overlaps with that of antibody clone CY1G4 but is distinct from the ligand binding site for transferrin.
Type:
Antigen: CD71
Clonality: Monoclonal
Clone: OKT9
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human