Order Entry
Denmark
ContactUsLinkComponent
Anti-Igm Isotype Control Rat Monoclonal Antibody [clone: RTK2118] (FITC)
Anti-Igm Isotype Control Rat Monoclonal Antibody [clone: RTK2118] (FITC)
Certificates
undefined
Anti-Igm Isotype Control Rat Monoclonal Antibody [clone: RTK2118] (FITC)

Specifications

  • Catalog No:
  • STMC60074FI
  • STMC60074FI.1
  • Antigen name:
  • Igm Isotype Control
  • Igm Isotype Control
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • RTK2118
  • RTK2118
  • Host:
  • Rat
  • Rat
  • Isotype:
  • IgM kappa
  • IgM kappa

Specifications

Frequently Bought Together