Order Entry
Denmark
ContactUsLinkComponent
Anti-CD150 (SLAM) Rat Monoclonal Antibody [clone: TC15-12F12.2] (APC)
Anti-CD150 (SLAM) Rat Monoclonal Antibody [clone: TC15-12F12.2] (APC)
Certificates
undefined
Anti-CD150 (SLAM) Rat Monoclonal Antibody [clone: TC15-12F12.2] (APC)

Specifications

  • Catalog No:
  • STMC60036AZ.1
  • STMC60036AZ
  • Antigen name:
  • CD150 (SLAM)
  • CD150 (SLAM)
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • TC15-12F12.2
  • TC15-12F12.2
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 27218
  • 27218
  • Isotype:
  • IgG2a lambda
  • IgG2a lambda

Specifications

Frequently Bought Together