Rat monoclonal IgG2a antibody against mouse Sca1 (Ly-6A/E), APC-conjugated.
- Hematopoietic stem
- Progenitor cells
- Mammary cells
The E13-161.7 antibody reacts with Sca1 (Stem Cell Antigen-1 or Ly-6A/E), an 18 kDa GPI-linked protein belonging to the lymphocyte activation protein-6 (Ly-6) family. Sca1 is expressed on the surface of hematopoietic stem and progenitor cells, myeloid cells, and peripheral B and T lymphocytes. Sca1 is expressed by mice with either the Ly-6.1 or Ly-6.2 allotypes but the pattern of expression differs in the circulating cell population according to the allotype. Ly-6.2 strains (e.g., AKR, C57BL, C57BR, C57L, DBA/2, PL, SJL, SWR, 129) possess relatively high numbers of Sca1+ resting lymphocytes compared to Ly-6.1 strains (e.g. A, BALB/c, CBA, C3H/He, DBA/1, NZB). Sca1 expression levels are strongly upregulated in all strains upon cellular activation. Sca1 is involved in the regulation of T and B cell responses and is believed to play roles in the differentiation, proliferation, and survival of a variety of stem cells. Sca1 has emerged as a phenotypic marker of choice for identifying and isolating hematopoietic stem and progenitor cells. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse SCA1 Biotin Positive Selection Kit (Catalog #18856).
Type:
Antigen: Sca1 (Ly-6A/E)
Clonality: Monoclonal
Clone: E13-161.7
Conjugation: APC
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
- Catalog No:
- STMC60032AZ
- STMC60032AZ.1
- Antigen name:
- Sca1 (Ly-6A/E)
- Sca1 (Ly-6A/E)
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- E13-161.7
- E13-161.7
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 110454
- 110454
- Isotype:
- IgG2a kappa
- IgG2a kappa