About this item
Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD8a, FITC-conjugated.
- T cells
- T cells CD8
The RPA-T8 antibody reacts with CD8a, a 32 - 34 kDa type I transmembrane glycoprotein which is a subunit of CD8. CD8 is a disulfide-bonded dimer, found either as a heterodimer of CD8a (α) and CD8b (β) subunits (i.e., αβ) or a homodimer (αα). CD8 acts as a co-receptor to the T cell receptor (TCR) during T cell activation by binding MHC Class I molecules presented by an antigen-presenting cell. It functions to strengthen the association between the TCR and MHC I-antigen complex, and to amplify signals from the TCR to the cytoplasm through the interaction of its intracellular domain with cytoplasmic tyrosine kinases such as Lck. The CD8a chain binds to the α3 domain of class I MHC molecules. CD8 is expressed by a majority of thymocytes, a subset of mature peripheral blood T cells (T cytotoxic cells), a proportion of β T cells, and at lower levels by NK cells (which predominantly express CD8a homodimers). This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Direct Human CD8+ T Cell Isolation Kit (Catalog #19663), EasySep™ Human CD8+ T Cell Enrichment Kit (Catalog #19053), EasySep™ Human CD8 Positive Selection Kit (Catalog #18053), EasySep™ Human CD3 Positive Selection Kit (Catalog #18051), EasySep™ HLA CD2 Positive Selection Kit (Catalog #18657HLA), and EasySep™ HLA T Cell Enrichment Kit (Catalog #19051HLA).
Type:
Antigen: CD8a
Clonality: Monoclonal
Clone: RPA-T8
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Baboon, Chimpanzee, Cynomolgus monkey, Human, Macaque, Rhesus monkey, Sooty mangabey
Specifications
- Catalog No:
- STMC60022FI
- STMC60022FI.1
- Antigen name:
- CD8a
- CD8a
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- RPA-T8
- RPA-T8
- Reactivity:
- Baboon, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Macaque, Chimpanzee, Human
- Baboon, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Macaque, Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 925
- 925
- Isotype:
- IgG1 kappa
- IgG1 kappa